Anti-Trem1 IgG Polyclonal Antibody

Supplier: Boster Biological Technology
A02135-2
76171-570EA 702.73 USD
76171-570
Anti-Trem1 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for TREM1 detection. Tested with WB, ELISA(Cap) in Mouse;Rat.

Type: Primary
Antigen: Trem1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR