Anti-NEDD4L IgG Polyclonal Antibody

Supplier: Boster Biological Technology
A01595-1
76171-472EA 787.77 USD
76171-472
Anti-NEDD4L IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for NEDD4-2 detection. Tested with WB, Direct ELISA in Human;Mouse;Rat.

Type: Primary
Antigen: NEDD4L
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR