Anti-BNIP3 IgG Polyclonal Antibody

Supplier: Boster Biological Technology
A01469
76171-446EA 702.73 USD
76171-446
Anti-BNIP3 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for BNIP3 detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: BNIP3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR