Microflex® 93-850 Ultimate Barrier Disposable Nitrile Gloves, Ansell
Supplier: Ansell Healthcare
93850070
76169-806BX 54.23 USD
76169-806 76169-808 76169-908 76169-910 76169-912 76169-914
Microflex® 93-850 Ultimate Barrier Disposable Nitrile Gloves, Ansell
Gloves
Ultimate barrier disposable nitrile glove with TNT™ technology.
  • 2X more chemical splash protection than leading brands due to breakthrough polymer bonding technique
  • Exceeds every known standard for barrier quality and consistency (Low 0.40 AQL)
  • Made with TNT™ Chemical Splash resistance technology, for soft durable protection against a wide range of chemicals
  • Silicone-free formulation for product protection
Microflex® 93-850 nitrile disposable gloves represent the ultimate barrier against harmful exposures. These tough, durable gloves stand up to tough jobs while significantly reducing workers’ risk of exposure to hazardous materials.
Caution: Based on EN 374 and ASTM F739 chemical permeation test data when compared to performance of disposable nitrile gloves of a similar weight. Based on Ansell durability pull test data, when compared to performance of disposable nitrile gloves of similar thickness. Microflex® 93-850 0.40 AQL is lower than the AQL of other known industrial and medical grade disposable gloves.