AlphaTec® 87-297 Natural Rubber Latex Gloves with Cotton Lining, Yellow, Ansell
Supplier: Ansell Healthcare
120193
76169-859CS 169.92 USD
76169-859 76169-861 76169-863 76169-865
AlphaTec® 87-297 Natural Rubber Latex Gloves with Cotton Lining, Yellow, Ansell
Gloves
Gloves combining strength, sensitivity and grip. Medium duty, is both flexible and comfortable.
  • Material provides a high degree of tensile strength
  • 100% cotton flock lining helps maintain excellent sensitivity
  • Provides snag, puncture and abrasion resistance
  • Embossed pebble and fish scale designs provide improved grip on wet and dry objects
Ideal applications include chemical handling, clean room cleaning and preparing, and re-fuel oil and diesel.
Certifications: ANSI Abrasion 3. EN ISO 374-1 Type C. EN 388:2016 - 1010X. EN 388:2003 - 2110.