Blood Agar, 5% Sheep in Tryptic Soy Agar (TSA) Base, with Ampicillin
Supplier: HARDY DIAGNOSTICS
A12
76169-804PK 54.56 USD
76169-804
Blood Agar, 5% Sheep in Tryptic Soy Agar (TSA) Base, with Ampicillin
Media for Microbiology Prepared Media for Microbiology
TSA with 5% Sheep Blood and Ampicillin is ideal for the selective isolation of Aeromonas spp.
This product has a short shelf life and ships from California only.