Innoculating Needle
Supplier: Hardy Diagnostics
HSTND
76169-774EA 195.07 USD
76169-774
Innoculating Needle
Spreaders Inoculating Loops and Needles
Hexagonal needle shaft improves grip and is free of lubricants, oils, and electrostatic charges.
Color coded for easy product identification.
Packaging: Packaged in rectangular, plastic tubes of 50 needles with lid that allows placement on clean table without rolling; 1000 needles per box.