HardyDisks™ Trimethoprim, TMP-5, AST
Supplier: HARDY DIAGNOSTICS
Z9181
76169-772EA 32.57 USD
76169-772
HardyDisks™ Trimethoprim, TMP-5, AST
Microbiology Test Systems Antibiotic Tests
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
  • Impregnated disks
  • Spring loaded dispenser
  • Smooth and reliable dispensing
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.