HardyDisks™Cephalothin, CF-30, AST, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z8331
76169-768EA 31.66 USD
76169-768
HardyDisks™Cephalothin, CF-30, AST, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing. The HardyDisk™ AST cartridges are designed to be fully compatible with the BBL single or multiple disk dispensers.