VWR® High Performance Multi-Channel Pipettors
Supplier: VWR International
76169-246 76169-248 76169-250 76169-252 76169-254 76169-256 76169-258 76169-260 76208-498 76208-502 76208-504
76169-246EA 824.37 USD
76169-246 76169-248 76169-250 76169-252 76169-254 76169-256 76169-258 76169-260 76208-498 76208-502 76208-504
VWR® High Performance Multi-Channel Pipettors
Pipettes Micropipettes
VWR® High Performance Pipettors are the newest lightweight pipettor designed for user comfort throughout daily work. Thanks to low pipetting forces, users get a unique pipetting experience. With its, high accuracy and precision, the VWR® High Performance Pipettor delivers highly precise and reproducible results.
  • Exclusive volume lock mechanism
  • Lightweight design
  • Low pipetting forces
  • Outstanding precision and accuracy
  • Fully autoclavable without disassembly (120 °C/0.1 MPa/20 min.) and UV resistant
  • Universal tip fitting
  • Unique serial number
VWR® High Performance Pipettors have been designed to fit both right- and left-handed users while providing maximum comfort.
To save time, the multi-channel pipettor mechanism offers the same performance on each channel by handling multiple samples consistently. The 360° rotation feature allows multiple pipetting orientations. VWR® High Performance Pipettors are available in a large volume range, with 8 multi-channel Pipettors from 0.5 to 300 µl (8 and 12 channels).
The large ejector button is built to fit either hand perfectly and allows pipetting without fatigue. A single button is used to eject and secure the volume setting. A unique instant volume locking system mechanism prevents accidental volume change while in use. These features combine to allow easy one-handed operation. Simply pull the ejection button, adjust the volume with the push button, and lock the pipettor by pushing down the ejection button. Digits are easy to read and adjust. The volume is always visible while pipetting. Easy to maintain, VWR® High Performance Pipettors are developed to simplify daily use thanks to intuitive in-lab disassembly, maintenance, and a high chemical compatibility.
Certifications: CE marked.
Ordering information: Each order includes a 36 month warranty. The product comes with a certificate of conformity, a calibration key, a Quick start guide, and a tip ejector extension (for 10 µl model only).