AimStrip® Urine Analyzer II, Germaine Laboratories

Supplier: Germaine Laboratories
51222
76169-158CS 0.02 USD
76169-158
AimStrip® Urine Analyzer II, Germaine Laboratories
Clinical Diagnostic Supplies Clinical Analysers
The AimStrip® Urine Analyzer 2 reads urine test strips, stores results and prints reports, all without the need for special training.

  • Single Test Mode, reads 1 strip at a time
  • Continuous Test Mode, reads 120 strips
  • Automatic calibration for accurate results
  • Automatically saves the last 2000 results
  • Reads 2, 3, 4, 5, 6, 7, 8, 9, and 10 parameter strips
  • Includes RS232C and USB ports to export results

The AimStrip® Urine Analyzer 2 is intended for use in conjunction with the AimStrip® Urinalysis Reagent Strips for the semi-quantitative detection of the following analytes in urine: Glucose, Bilirubin, Ketone (Acetoacetic acid), Specific Gravity, pH, Blood, Protein, Urobilinogen, Leukocytes, Ascorbic Acid, as well as the qualitative detection of Nitrite. The instrument is intended for professional, in vitro diagnostic use only. The measurement can be used in general evaluation of health, and aids in the diagnosis and monitoring of metabolic or systemic diseases that affect kidney function, endocrine disorders and diseases or disorders of the urinary tract.

Additional information
Weight: 8 lbs
Dimensions: 19.25 × 11 × 9 in
Electrical: 100 - 240V AC, 50/60Hz, 35 VA

Certifications: CLIA Waived. This product complies with EN 61326.

Accessories information: Barcode Reader Optional.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR