Jenway® 72 Series UV/Visible Diode Array Scanning Spectrophotometer, Model 7206 Genova Bio, Cole-Parmer
Supplier: Antyila Scientific
83056-04
76169-036EA 7144.68 USD
76169-036
Jenway® 72 Series UV/Visible Diode Array Scanning Spectrophotometer, Model 7206 Genova Bio, Cole-Parmer
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Diode array technology speeds up spectral data acquisition.
  • Simultaneous measurement of all wavelengths increases productivity, scan time as short as 3 seconds
  • Multiple measurement modes for broad application compatibility
  • Color touch screen and user-friendly interface make setup easy
  • Preprogrammed for DNA, RNA and protein analysis
  • Simple and easy-to-use at an affordable price
  • Narrow spectral bandwidth of 3 nm identifies peaks for determining nucleic acid purity
  • Dedicated to life science applications
The multi-element detectors on these scanning spectrophotometers measures the entire wavelength range at once, reducing wait times. Plus, each dataset contains readings from the complete spectrum, making the data readily available for future analysis. The detector's orientation shields the array from stray light, allowing you to perform measurements with the lid open, quickly exchange samples, and decreases user errors in results.
Small footprint takes up less bench space, the large color touch screen is easy to read, and the graphical interface is intuitive. The USB port on the rear connects with a printer (order separately), while the USB on the front accepts flash drives for data storage, quantitation curve uploads, and firmware updates.
Measurement modes include photometrics, concentration, optical density (Ext.Ø at 600 nm), quantitation, spectrum scanning, and kinetics.