Distillation Rods, Büchi
Supplier: Buchi
043087
76169-032EA 1792.29 USD
76169-032
Distillation Rods, Büchi
Solid Rods
Reusable glass rods designed specifically to aid in digestion of samples for Kjeldahl determination.
  • Prevent bumping
  • Minimize foaming
  • Designed not to interfere with aspiration of automated systems
Aids in minimizing foaming of high surface tension samples to keep entire sample contents within the reflux zone. Helps to equilibrate heat evenly throughout the samples.