Kjeldahl Weighing Boats, N-free, Buchi
Supplier: Buchi
11060522
76169-030EA 418.74 USD
76169-030
Kjeldahl Weighing Boats, N-free, Buchi
Boats Weighing Boats
Weighing boats for powder, solids or viscous fluids for reliable nitrogen and protein determination according to Kjeldahl.
  • Convenient handling and easy transfer into sample tube
  • Reliable results thanks to high quality
  • Nitrogen-free paper
Simply tare the boat, weigh and record sample, place boat into tube and raise tube to slide boat to the bottom of the vessel. Add acid and Kjeldahl tablet catalyst.