Rhodium 5% on activated carbon powder wetted with water 27 µm for use as a catalyst, reduced , BASF Catalyst

Supplier: Spectrum Chemical Mfg. Corp.

SP-300-10G SP-300-1G SP-300-50G SP-300-5G
76124-508EA 488 USD
76124-508 76124-510 76124-512 76124-514
Rhodium 5% on activated carbon powder wetted with water 27 µm for use as a catalyst, reduced , BASF Catalyst
Rhodium

5 Percent Rhodium on Activated Carbon Powder, Reduced, Water Wet, 27 microns, BASF Catalyst

Formula: Rh
Storage Temperature: Ambient
MDL Number: MFCD00011201
CAS Number: 7440-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR