Palladium 3% on activated carbon powder wetted with water 30 µm for use as a catalyst, unreduced , BASF Catalyst

Supplier: Spectrum Chemical Mfg. Corp.

SP-104-100G SP-104-25G SP-104-5G
76124-436EA 486.3 USD
76124-436 76124-438 76124-440
Palladium 3% on activated carbon powder wetted with water 30 µm for use as a catalyst, unreduced , BASF Catalyst
Palladium

3 Percent Palladium on Activated Carbon Powder, Unreduced, Water Wet, 30 microns, BASF Catalyst


Caution: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Formula: Pd
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR