HardyVal™ MTK (Multiple Technician Kit)
Supplier: HARDY DIAGNOSTICS
HVMTK
76121-434EA 172.6 USD
76121-434
HardyVal™ MTK (Multiple Technician Kit)
Microbiology Test Systems Microorganism Tests
HardyVal™ MTK (Multiple Technician Verification Kit) is recommended for verification of personal aseptic technique for low and medium complexity levels within a sterile compounding pharmacy facility or other cleanroom application.
Each kit contains enough media to perform aseptic technique verification for up to five technicians. Product is quality control tested for growth promotion, pH, and sterility.
Delivery information: Each kit contains 5 ampoules of TSB, 3 ml (non-toxic red dye is added to the TSB to easily visualize the sample being transferred), 5 vials of TSB 20 ml serum needle-port septum vial, 5 bags of TSB 100 ml media bag.