Mini Centrifuge, BioAssay Systems
Supplier: BIOASSAY SYSTEMS
CTFG-100
76121-354EA 80.1 USD
76121-354
Mini Centrifuge, BioAssay Systems
Centrifuges Benchtop Centrifuges
Small Centrifuge used for preparation of fish and wine samples for use with the QuantiQuik™ Histamine Quick Test Strips