Histamine Quick Test Strips, BioAssay Systems
Supplier: BIOASSAY SYSTEMS
QQHIST10
76121-346EA 111.79 USD
76121-346
Histamine Quick Test Strips, BioAssay Systems
Assays Cellular Assays
Direct determination of Histamine concentrations in fish and red wine samples.
  • Fast and sensitive. Use of 500 mg of fish or 250 µL of sample.
  • Semi-quantitative measurement between 0-200 ppm (undiluted) Histamine.
  • Convenient.
  • Sample treatment and assay can be performed in under 15 minutes.
  • Method: Visual
  • Samples: Seafood, wine, milk, etc
HISTAMINE(C5H9N3) is a biogenic amine naturally present in many foods and body cells in amounts without toxicological significance. It is also a contaminant mostly found in Scombridae family fishes, such as tuna and mackerel, as well as other seafood products when they are improperly handled or stored. The consumption of foods containing high levels of histamine may lead to an allergy-like food poisoning known as scombroid poisoning. BioAssay Systems' QuantiQuik™ Histamine Quick Test Strips are based on histamine dehydrogenase catalyzed oxidation of histamine in which the formed electron mediator reduces a formazan (MTT) reagent. The intensity of product color is directly proportional to histamine concentration in the sample.