Anti-ELYS Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss

BS-9880R-A750
76120-276EA 732.61 USD
76120-276
Anti-ELYS Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
ELYS is a protein of the nuclear matrix that contains one AT hook DNA-binding domain. ELYS is a dual nucleoporin/kinetochore protein required for nuclear pore assembly and proper cell division. Nuclear pores span the nuclear envelope and act as gated aqueous channels to regulate the transport of macromolecules between the nucleus and cytoplasm, from individual proteins and RNAs to entire viral genomes. The A.T. hook near the C-terminus of likely indicates ELYS can function as a transcription factor which may play a specific role in hematopoietic tissues. It also may play a role in early embryo development, as ELYS is critical for the survival of inner cells of the blastocyst.

Type: Primary
Antigen: ELYS
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 451-550/2266
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR