Anti-SUGT1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss

BS-7831R-A680
76119-848EA 732.61 USD
76119-848
Anti-SUGT1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
May play a role in ubiquitination and subsequent proteasomal degradation of target proteins.

Type: Primary
Antigen: SUGT1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 271-365/365
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR