Anti-IFN-Alpha Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss

BS-7021R-A680
76119-092EA 732.61 USD
76119-092
Anti-IFN-Alpha Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Interferon-_ (IFN-_, also known as IFN-alpha, IFN-a, IFNA and Interferon-alpha) is a type I interferon and exists as 13 subtypes: IFN-_ 1, 2, 4, 5, 6, 7, 8, 10, 13, 14, 16, 17 and 21. They are mainly produced by virus-infected cells after sensing the presence of danger signals such as viral DNA or RNA. The binding of IFN-_ to its receptor initiates an anti-viral response, but also anti-tumor effects have been shown. A key producer of type I interferons is the plasmacytoid dendritic cell.

Type: Primary
Antigen: IFN-Alpha
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-189/189
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR