Anti-BRF1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss

BS-6358R-A680
76116-262EA 732.61 USD
76116-262
Anti-BRF1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
General activator of RNA polymerase which utilizes different TFIIIB complexes at structurally distinct promoters. The isoform 1 is involved in the transcription of tRNA, adenovirus VA1, 7SL and 5S RNA. Isoform 2 is required for transcription of the U6 promoter.

Type: Primary
Antigen: BRF1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 501-600/677
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR