Anti-AGFG1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss

BS-8072R-A750
76108-220EA 732.61 USD
76108-220
Anti-AGFG1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Required for vesicle docking or fusion during acrosome biogenesis (By similarity). May play a role in RNA trafficking or localization. In case of infection by HIV-1, acts as a cofactor for viral Rev and promotes movement of Rev-responsive element-containing RNAs from the nuclear periphery to the cytoplasm. This step is essential for HIV-1 replication.

Type: Primary
Antigen: AGFG1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 276-380/562
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR