Face Shield Plus, PW96, Portwest
Supplier: Portwest
PW96CLR
76105-172EA 23.36 USD
76105-172
Face Shield Plus, PW96, Portwest
Face Shields
This face shield protects against molten metal splashes and hot solid projections. Superior front and side face protection. Ratchet adjustment for comfort fit.
  • Browguard including clear polycarbonate visor with aluminum edge
  • Protects from impacts, metal splashes and hot solid projections
  • Suitable for a wide range of applications
  • Wheel ratchet size adjustment for easy fitting
  • Simple and secure visor replacement
Replacement visor available (PW99).
Certifications: CE certified. Meet EN 166 (1 BT 3 9), ANSI/ISEA Z87 +