Reinforced Welding Gauntlet, Portwest
Supplier: Portwest
A530BRRXL
76103-626EA 17.47 USD
76103-626
Reinforced Welding Gauntlet, Portwest
Gloves
The reinforced welding gauntlet is made from premium quality leather with reinforced palm and thumb area.
  • Fully welted and sewn with para-aramid throughout
  • EN12477 Type A welding protection
  • Reinforced palm and thumb area
  • Para-aramid stitching throughout for extra durability
  • Superb abrasion and tear resistance
Ideal for welding and metal handling.
Certifications: AS/NZS 2161 .2. AS/NZS 2161 .3. EN 407. EN 420. EN 388:2003 - 3.1.4.4. EN 12477 Type A. ANSI/ISEA 105 - 2011 Abrasion Level (4).