Welders Gauntlet, Portwest
Supplier: Portwest
A500RERXL
76103-624EA 12.69 USD
76103-624
Welders Gauntlet, Portwest
Gloves
The tough 14" split leather gauntlet providing protection against a range of heat related tasks. Ideal for welding and metal handling due to the premium quality materials used.
  • EN12477 Type A welding protection
  • Fully lined with soft and comfortable cotton
  • 14 inch cow split leather gauntlet
  • Maximum EN407 burn behaviour resistance
  • Durable leather
Ideal for welding and metal handling.
Certifications: AS/NZS 2161 .2. AS/NZS 2161 .3. AS/NZS 2161 .4. ANSI/ISEA 105 - 2016 ABRASION Level (3). EN420. EN388:2003 - 3.2.3.3. EN407 4.2.3.X.3.X. heat and fire protection. EN12477 Type A.