Dexti-Grip Gloves, Portwest
Supplier: Portwest
A320BKRL A320BKRM A320BKRXL A320BKRXXL
76103-574EA 1.68 USD
76103-574 76103-576 76103-578 76103-580
Dexti-Grip Gloves, Portwest
Gloves
Designed for intricate tasks. This glove is the popular choice for general assembly work.
  • Nitrile foam coating for excellent grip in wet and dry conditions
  • Palm dipped to increase dexterity and ventilation
  • 100% breathable seamless pylon liner
Certifications: AS/NZS 2161 .2. AS/NZS 2161 .3. ANSI/ISEA 105 - 2016 ABRASION Level (3). EN 388:2003 - 3.1.3.1. EN 420.