HBS-EP+ buffer 20X, sterile, Teknova
Supplier: TEKNOVA INC
H8022
76102-774EA 182.14 USD
76102-774
HBS-EP+ buffer 20X, sterile, Teknova
Buffers Electrophoresis Buffers Running Buffers
Ingredients: 0.2 M hepes, 3 M sodium chloride, 60 mM EDTA, 1.0% polysorbate 20, pH 7.6.