Human Fibronectin (from Plasma), MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0215112601
76102-632EA 395.04 USD
76102-632
Human Fibronectin (from Plasma), MP Biomedicals
Proteins and Peptides
Fibronectins are adhesive glycoproteins and are found both as cell surface proteins and in plasma.
  • Suitable for cell culture
  • Presentation: White Solid
  • Soluble in water (1 mg/mL at 37 °C).
Human fibronectin (HFN) is suitable for use as an attachment factor in the propagation of cells in vitro when used to coat cell culture surfaces, including plasticware, glassware, and microcarrier beads. It is used in cell attachment and proliferation assays on human endothelial cells, human keratinocytes and human dermal fibroblasts.
HFN is purified from Cohn fraction I of human plasma by a modification of the procedure. This material has been pasteurized to reduce the possibility of viral contamination, and filtered through a 0.22 micron filter prior to lyophilization.
Reconstitution: Reconstitute in physiological buffer or urea for best results. After reconstitution, store in convenient aliquots at -20 °C for up to one year. Avoid repeated freeze/thaw cycles.
HFN is purified from Cohn fraction I of human plasma by a modification of the procedure. This material has been pasteurized to reduce the possibility of viral contamination, and filtered through a 0.22 micron filter prior to lyophilization.