Alcian blue 8GX, dark blue powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215262405
76102-598EA 128.77 USD
76102-598
Alcian blue 8GX, dark blue powder
Alcian blue 8 GX

Alcian Blue 8GX is a cationic dye used to quantitatively determine glycosaminoglycans by forming insoluble complexes in solution.


Alcian Blue 8GX can be used to determine proteins based on their Resonance Light Scattering by strongly enhancing the signals. It is useful as a bacterial stain, and as a dye for histiocytes and fibroblasts. Alcian Blue 8GX is used as a heteroglycan stain for neutral, sulfated and phosphated mucopolysaccharides and glycosaminoglycans in tissues such as cartilage and extracellular matrices.


Soluble in absolute ethanol, cellosolve, ethylene glycol, water (8 mg/ml) or ethanol (1 mg/ml); practically insoluble in xylene


Caution: Research Use Only (RUO).

Formula: C₅₆H₆₈Cl₄CuN₁₆S₄
Storage Temperature: Ambient
MDL Number: MFCD00010720
CAS Number: 33864-99-2

Order Now

Specification Test Results

Identity Test Passes
UV/VIS: λmax (water) 620 - 630 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR