NOVA-TECH 10 BLUE™ Non-Woven Cleanroom Wipes, High-Tech Conversions
Supplier: High-Tech Conversions
NT1B-99
76102-176BG 27.3 USD
76102-176
NOVA-TECH 10 BLUE™ Non-Woven Cleanroom Wipes, High-Tech Conversions
Wipes Cleanroom Wipes Dry Cleanroom Wipes
NOVA-TECH 10 BLUE™ is a lightweight, low-linting, poly-cellulose cleanroom wipe.
  • Class 100 cleanroom or higher
  • Autoclavable
  • Tough and absorbent
  • Virtually free of particle generation
NOVA-TECH 10 BLUE™ is an especially effective low-linting wipe.
These unique features give NOVA-TECH 10 BLUE™ a high degree of absorbency and effectiveness during the process of general wiping. Wipes made with this fabric are converted in a class 100 cleanroom.
The wipe is comprised of a synergistic blend of virgin polyester and cellulose that is hydroentangled into a uniform, super-tough fabric.
Packaging: Wipes are packaged 300 wipes per bag, 14 bags per case.