Anti-alpha Adducin Ser436 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)

Supplier: Bioss

BS-15968R-A750
76100-500EA 753.8 USD
76100-500
Anti-alpha Adducin Ser436 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Antibodies
Adducins are a family of cytoskeleton proteins encoded by three genes (alpha, beta, gamma). Adducin is a protein associated with the inner leaflet of the plasma membrane and is one of the proteins localized at the spectrin-actin junction of the membrane skeleton. The cortical actin cytoskeletal network is lost during apoptosis and Adducins are central in the cortical actin network organization. Adducin Alpha is a cytoskeletal protein involved with sodium-pump activity in the renal tubule and is associated with hypertension. The expression of Adducin Alpha and Adducin Gamma is ubiquitous in contrast to the restricted expression of Adducin Beta. Adducin Beta is expressed at high levels in brain and hematopoietic tissues, such as bone marrow, in humans, and in spleen in mice.

Type: Primary
Antigen: alpha Adducin Ser436
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR