Anti-E.coli DH-5 Alpha Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss

BS-2033R-A680
76098-586EA 753.78 USD
76098-586
Anti-E.coli DH-5 Alpha Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
This antibody reacts with Escherichia coli HD5a (E.coli HD5a), Escherichia coli is a gram-negative bacillus that belongs to a larger group of Enterobacteriae - bacteria that inhabit the gastrointestinal tract. Although usually a harmless resident of the gut, some strains have the potential to cause serious problems, especially where there is an immature immune system or immunosuppression, or where the subtype of organism has acquired the ability to produce pathogenic toxins. E. coli DH5_ strain is an engineering strain widely used in biopharmaceuticals. According to the requirements of product quality inspection, it is necessary to detect the residual amount of DH5_ bacterial protein in biological products. This product is an anti-serum obtained by immunizing rabbits with DH5_ bacterial protein as an antigen, and purified by affinity chromatography to obtain antibody IgG, which can be used for ELISA, Western Blot and other experimental tests.

Type: Primary
Antigen: E.coli DH-5 Alpha
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria, Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR