Anti-TLK1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss

BS-3583R-A750
76084-446EA 753.79 USD
76084-446
Anti-TLK1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
TLK1 is a nuclear serine/threonine kinases that are potentially involved in the regulation of chromatin assembly. TLK1 is rapidly and transiently inhibited by phosphorylation following the generation of DNA double-stranded breaks during S-phase. This is cell cycle checkpoint and ATM-pathway dependent and appears to regulate processes involved in chromatin assembly. Isoform 3 phosphorylates and enhances the stability of the t-SNARE SNAP23, augmenting its assembly with syntaxin.

Type: Primary
Antigen: TLK1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 458-560/766
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR