QuickExtract™ Plant DNA Extraction Solution, PCR-ready DNA from Leaves, Biosearch Technologies
Supplier: Biosearch Technologies
QEP70750
76081-772EA 743.21 USD
76081-772
QuickExtract™ Plant DNA Extraction Solution, PCR-ready DNA from Leaves, Biosearch Technologies
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Rapid and efficient extraction (no purification) of PCR-ready genomic DNA from plant leaves
  • Fast: PCR-ready DNA in about 8 minutes for most samples
  • Simple: No bead-beating, freezing, or grinding of plant leaf material
  • Increased Yields: No centrifugation or spin columns used which reduce yields
  • Compatible: Extracted DNA is compatible with both real-time and endpoint PCR
  • Safe: Nontoxic reagents used throughout the procedure
The QuickExtract™ Plant DNA Extraction Solution can be used to rapidly and efficiently extract PCR-ready genomic DNA from most plant leaf samples using a simple, one-tube protocol that takes only 8 minutes
The QuickExtract Plant method allows for the inexpensive processing of one to hundreds of samples simultaneously, without grinding the sample, centrifugation, spin columns, or any toxic organic solvent. The procedure is fully compatible with robotic automation, provides a PCR-ready sample, and is reproducible. Simply add the QuickExtract Plant solution to the sample and perform two sequential heating steps. A small aliquot of the sample is then used as a template for PCR or qPCR.
Most leafy plants are suitable for DNA extraction using the QuickExtract Plant Solution, including Arabidopsis, barley, maize, emmer, pepper, rice, spelt, spinach, soybeans, and wheat.