MasterPure™ Gram Positive DNA Purification Kit, Biosearch Technologies
Supplier: Biosearch Technologies
MGP04100
76081-754EA 744.46 USD
76081-754
MasterPure™ Gram Positive DNA Purification Kit, Biosearch Technologies
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Purify genomic DNA from challenging Gram positive bacteria for a wide variety of molecular biology applications
The MasterPure™ Gram Positive DNA Purification Kit provides all of the reagents needed to purify DNA from Gram positive bacteria
Examples of Gram-positive bacteria tested with this kit include Bacillus subtilis, Listeria monocytogenes, Staphylococcus aureus, Streptococcus mutans, and Lactococcus lactis. The kit effectively purifies high molecular weight genomic DNA which is suitable for PCR analysis and many other molecular biology applications.
Produces clean, high molecular weight genomic DNA ready for PCR and other molecular biology applications such as restriction digestion, cloning, and Southern blotting
Scaleable method for large sample volumes with no wasting of reagents with smaller samples
Complete kit includes lysozyme and does not require any additional purchases
Flexible kit also purifies gDNA from Gram negative bacteria
Robust kit works on a variety of tested species
These bacteria lyse more readily after treatment with Ready-Lyse™ Lysozyme and the Gram-Positive Cell Lysis Solution included in the kit. Ready-Lyse Lysozyme is a stable solution of a recombinant lysozyme from a nonmammalian, nonavian source. It has high specific activity and does not bind DNA.