Exo-Minus Klenow DNA Polymerase (D355A, E357A), Complete Exonuclease Minus, Biosearch Technologies
Supplier: Biosearch Technologies
KL11101K
76081-728EA 535.03 USD
76081-728
Exo-Minus Klenow DNA Polymerase (D355A, E357A), Complete Exonuclease Minus, Biosearch Technologies
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
An improved Klenow DNA polymerase lacking all 5' to 3' and 3' to 5' exonuclease activity
  • Synthesize DNA from ssDNA templates without degrading any single-stranded DNA
  • Suitable for a variety of applications including random-primer labeling of DNA, second-strand cDNA synthesis, strand-displacement amplification
Exo-Minus Klenow DNA Polymerase is a DNA-dependent DNA polymerase that lacks both the 5´ to 3´ and 3´ to 5´ exonuclease activities of E. coli DNA Polymerase I, from which it is derived. This N-terminal truncation of DNA Polymerase I also has two mutations (D355A and E357A).
Certifications: For research use only. Not for human or diagnostic use.
Ordering information: Contents include enzyme and a 10X reaction buffer.