Endonuclease
Supplier: Biosearch Technologies
OC7850K
76081-706EA 716.32 USD
76081-706
Endonuclease
Enzymes
Remove all types of RNA and DNA with this robust, flexible nuclease
  • Non-specific: Digests all types of DNA and RNA including single- or double-stranded linear, circular, and supercoiled
  • Flexible: Active within a broad range of conditions normally used in protein purification
  • Customizable: Contact us for bulk or high concentration enzyme
OmniCleave™ Endonuclease is a highly purified enzyme from a recombinant E
Applications include:
Removal of nucleic acids from cell lysates (reduction of viscosity) for improved handling and yield of protein preparations
Removal of trace contamination by nucleic acids in protein preparations
Removal of host DNA from phage preparations
Improved electrophoretic and chromatographic separation of proteins isolated from whole-cell lysates
coli strain that degrades single- and double-stranded DNA and RNA to di-, tri-, and tetranucleotides. OmniCleave Endonuclease has the same substrate specificity and yields the same products as Benzonase®, an enzyme derived from Serratia marcescens.
Certifications: For research use only. Not for human or diagnostic use.
Ordering information: Contents include enzyme and a dilution buffer.