Anti-KLST Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss

BS-5786R-A750
76081-188EA 736.09 USD
76081-188
Anti-KLST Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Kallistatin is a serine proteinase inhibitor which binds to tissue kallikrein and inhibits its amidolytic and kininogenase activity. Inhibition is achieved by formation of an equimolar, heat- and SDS-stable complex between the inhibitor and the enzyme, and generation of a small C-terminal fragment of the inhibitor due to cleavage at the reactive site by tissue kallikrein. Heparin blocks kallistatin's complex formation with tissue kallikrein and abolishes its inhibitory effect on tissue kallikrein's activity.

Type: Primary
Antigen: KLST
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 151-250/427
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR