Anti-NNT Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss

BS-5097R-A750
76079-716EA 753.79 USD
76079-716
Anti-NNT Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
The transhydrogenation between NADH and NADP is coupled to respiration and ATP hydrolysis and functions as a proton pump across the membrane. May play a role in reactive oxygen species (ROS) detoxification in the adrenal gland.

Type: Primary
Antigen: NNT
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 981-1086/1086
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR