Anti-Hepatitis C Virus RNA-directed RNA polymerase Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss

BS-4858R-A680
76079-254EA 753.79 USD
76079-254
Anti-Hepatitis C Virus RNA-directed RNA polymerase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
The RNA directed RNA polymerase is also known as non-structural protein NS5B. NS5B is a 65 kDa protein that resembles other viral RNA polymerases. Hepatitis C virus (HCV) replication is thought to occur in membrane bound replication complexes. These complexes transcribe the positive strand and the resulting minus strand is used as a template for the synthesis of genomic RNA. There are two viral proteins involved in the reaction, NS3 and NS5B.

Type: Primary
Antigen: Hepatitis C Virus RNA-directed RNA polymerase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 2501-2600/3010
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR