Anti-Nonstructural Protein 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss

BS-4839R-A750
76079-232EA 753.79 USD
76079-232
Anti-Nonstructural Protein 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Influenza non-structural protein (NS1) is a homodimeric RNA-binding protein found in influenza virus that is required for viral replication. NS1 binds polyA tails of mRNA, keeping them in the nucleus.

Type: Primary
Antigen: Nonstructural Protein 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR