Frosted Slides, Azer Scientific
Supplier: Azer Scientific Inc.
231151A
76076-738CS 716.14 USD
76076-738
Frosted Slides, Azer Scientific
Slides Microscope Slides
Smooth slides with burr-free, wet-ground edges are frosted at a single end in order to prevent jamming and sticking.
Packaging: Slides are packaged 72 per box, 20 boxes per case.