Methanol for QuickSlide instrumentation

Supplier: HARDY DIAGNOSTICS

QS1016
76076-692EA 18.84 USD
76076-692
Methanol for QuickSlide instrumentation
Methanol

Designed for use in the fixation of specimens before Gram staining.


Caution: Note: This product is hazardous and ships ground only.

Formula: CH₃OH
MDL Number: MFCD00004595
CAS Number: 67-56-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR