Immersion Oil, Type A, Low Viscosity, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z95
76076-690EA 46.84 USD
76076-690
Immersion Oil, Type A, Low Viscosity, Hardy Diagnostics
Stains Microscopy and staining reagents
Used to assist in microscopic examination at high power (1,000 X lens) by increasing the resolving power
Type A is a low viscosity Immersion Oil.