Anti-SLC22A4 Rabbit Polyclonal Antibody

Supplier: Prosci
62-124
76072-192EA 636.42 USD
76072-192
Anti-SLC22A4 Rabbit Polyclonal Antibody
Antibodies
For WB starting dilution is: 1:1000

Type: Primary
Antigen: SLC22A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 514-542
Host: Rabbit
Isotype: Ig
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR