Anti-PAQR7 Rabbit Polyclonal Antibody

Supplier: Prosci
58-382
76069-030EA 617.88 USD
76069-030
Anti-PAQR7 Rabbit Polyclonal Antibody
Antibodies
For WB starting dilution is: 1:1000

Type: Primary
Antigen: PAQR7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 281-310
Host: Rabbit
Isotype: Ig
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR