Anti-OR2AG2 Rabbit Polyclonal Antibody

Supplier: Prosci
59-790
76063-954EA 617.88 USD
76063-954
Anti-OR2AG2 Rabbit Polyclonal Antibody
Antibodies
For WB starting dilution is: 1:1000

Type: Primary
Antigen: OR2AG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 64-90
Host: Rabbit
Isotype: Ig
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR