Anti-PACRG Rabbit Polyclonal Antibody

Supplier: Prosci
59-356
76063-230EA 617.88 USD
76063-230
Anti-PACRG Rabbit Polyclonal Antibody
Antibodies
For WB starting dilution is: 1:1000

Type: Primary
Antigen: PACRG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 46-74
Host: Rabbit
Isotype: Ig
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR